
Phenol red sodium salt, powder, Bioreagent cell culture reagent, Sigma-Aldrich®
Supplier: SIGMA ALDRICH MICROSCOPY
Certificates
About this item
CAS Number:34487-61-1
:
Formula:C₁₉H₁₃NaO₅S
MDL Number:MFCD00066901
:376.35 g/mol
Merck Index:12,07397
Melting Pt:285 °C (dec., lit.)
:M-Clarity™ quality level = MQ200
EINECS:252-057-8
:GHS07
Storage Temperature:Room Temperature
Specification Test Results
- Appearance (Colour):Red to Very Dark Red and Red-Brown and Brown-Red
- Appearance (Form):Powder
- Infrared spectrum:Conforms to Structure
- Solubility (Turbidity), 1 mg/ml, H₂O:Clear
- Suitability, Suitable for use as a pH indicator:Pass
- Visual Interval, pH 6.8 Yellow:Pass
- Visual Interval, pH 7.0 Orange:Pass
- Visual Interval, pH 8.2 Red:Pass
- Cell Culture Test:Pass
- Insect Cell Test:Pass
- Recommended Retest Period:5 Years
Specification Test Results
Frequently Bought Together