Order Entry
Switzerland
ContactUsLinkComponent
Phenolphthalein 0.1% in aqueous solution
Phenolphthalein 0.1% in aqueous solution
Catalog # HACH189736
Supplier:  Hach
CAS Number:  
undefined
Phenolphthalein 0.1% in aqueous solution
Catalog # HACH189736
Supplier:  Hach
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    15 mL
  • Seq:
    2295

Specifications

About this item

 :
 :318.33 g/mol
Storage Temperature:Ambient
EINECS:201-004-7
CAS Number:77-09-8
Formula:C₂₀H₁₄O₄
MDL Number:MFCD00005913