Pesticide reference standard, Chlorpyrifos 100 µg/ml in cyclohexane, Dr Ehrenstorfer
Catalog # EHERXA11600000CY
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 mL
- CAS No.:2921-88-2
- Description:Chlorpyrifos 100 µg/ml in cyclohexane
- Formula:C₉H₁₁Cl₃NO₃PS
- Description:Chlorpyrifos