Order Entry
Switzerland
ContactUsLinkComponent
Bromophenol blue sodium salt ACS
Bromophenol blue sodium salt ACS
Catalog # 0312-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Bromophenol blue sodium salt ACS
Catalog # 0312-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Clarity of Solution:
    Pass
  • Visual Transition Interval (pH 3.0 – 4.6):
    Pass

Specification Test Results

About this item

 :
 :691.95 g/mol
Storage Temperature:Ambient
EINECS:252-170-2
CAS Number:62625-28-9
Formula:C₁₉H₉Br₄NaO₅S
MDL Number:MFCD00013793