Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl red

Methyl red

Marketsource
Supplier:   Roth Carl
Methyl red
Supplier:  Roth Carl
Marketsource
Certificates

2 Options Available Below

Product Details & Documents
Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:12,06199
  • Melting Pt:178…182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1

Documents

Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - SFr. 27.00
    SFr. 27.00
    1 * 10 g
       SFr. 27.00

    Specifications:
    Marketsource
    Marketsource
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 43.10
    SFr. 43.10
    1 * 25 g
       SFr. 43.10

    Specifications:
    Marketsource
    Marketsource