Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Methyl red
Methyl red
Certificates
Methyl red

3 Options Available Below

Product Details & Documents

CAS No.: 493-52-7 Molecular Formula: C15H15N3O2 Formula Weight: 269.30 Melting Point: 179-182° Beilstein Reference: 750102 Merck Reference: 14,6119 MDL No.: MFCD00002425

Synonyms:  2-(4-Dimethylaminophenylazo)benzoic acid, 2-((4-(Dimethylamino)phenyl)diazenyl)benzoic acid
  • CAS Number:493-52-7
  • Formula:C₁₅H₁₅N₃O₂
  • MDL Number:MFCD00002425
  • Molecular Weight:269.3 g/mol
  • Merck Index:14,6119
  • Melting Pt:179…182 °C
  • Density:1.31 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:207-776-1
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - SFr. 51.50
    SFr. 51.50
    1 * 25 g
       SFr. 51.50

  • Glass bottle for solids
    100 g
    Log in for availability
    Log in for availability
    1 * 100 g
    1 * 100 g - SFr. 101.00
    SFr. 101.00
    1 * 100 g
       SFr. 101.00

  • Glass bottle for solids
    500 g
    Log in for availability
    Log in for availability
    1 * 500 g
    1 * 500 g - SFr. 321.00
    SFr. 321.00
    1 * 500 g
       SFr. 321.00