Order Entry
Switzerland
ContactUsLinkComponent
IL-1β (Human), ELISA Kit
IL-1β (Human), ELISA Kit
Catalog # ENZOADI900130A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
IL-1β (Human), ELISA Kit
Catalog # ENZOADI900130A
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 KIT
  • Primary antibody reactivity:
    Human
  • Target protein:
    IL-1β
  • Description:
    IL-1β (human), ELISA kit, 96 wells
  • Sensitivity:
    1 pg/ml

Specifications

About this item

For the quantitative determination of IL-1β in culture supernatant, plasma, serum, and urine of human origin.

  • Sensitive - quantify down to 1 pg/ml
  • Efficient - only 50 µl diluted sample required saving precious materials
  • Specific - no cross-reactivity with related cytokines
  • Results from up to 40 samples in duplicate in just 3 h
  • Ready-to-use reagents and pre-coated plates reduce errors and save your time

The IL-1β (human), EIA kit is a colorimetric immunometric enzyme immunoassay kit with results in less than 3 h. Absorbance is read at 450 nm. Quantify 1 pg/ml IL-1β with lot sample volume needed saving you precious sample.

Ordering information: Contains standard, standard diluent buffer, coated 96-well plate, conjugate, streptavidin-HRP, streptavidin-HRP diluent, wash buffer concentrate, TMB solution, stop solution, plate sealers.