5-Methylcytosine DNA ELISA Kit
Catalog # ENZOADI900224A0001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:1 KIT
- Primary antibody reactivity:Species independent
- Description:5-Methylcytosine DNA ELISA kit
- Detection method:Colorimetric 450 nm
- Storage temperature:4 °C (long term), −20 °C (standard and conjugate)
- Sensitivity:~0,5% 5-methylcytosine/100 ng single-stranded DNA (range 5 - 100% (100 ng/µl))
Specifications
About this item
The 5-Methylcytosine DNA ELISA kit is a complete, colorimetric, indirect immunoassay kit for the quantitative determination of 5-methylcytosine in DNA with results in less than three hours.
- Rapid – fully quantitative results in <3 hours
- Convenient – user-friendly alternative to mass spectrometry
- Simple – efficient and well-established protocol
- High-throughput – 96 well format ideal for global 5-methylcytosine detection in DNA
The ability to efficiently detect and quantify DNA methylation (i.e., 5-methylcytosine) has become essential for epigenetic-based research. To date, a number of methods have been developed for this purpose including high-performance capillary electrophoresis, bisulfite sequencing, and methylated DNA immunoprecipitation.
Shipping conditions: Dry ice