Hydroxystilbamidine bis(methanesulphonate) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52253
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:472.54 g/mol
Storage Temperature:Freezer
CAS Number:223769-64-0
Formula:C₁₈H₂₄N₄O₇S₂
MDL Number:MFCD01863083