Order Entry
Switzerland
ContactUsLinkComponent
Hygromycin B 100 mg/ml in water (from Streptomyces hygroscopicus) , 90% (by HPLC), sterile filtered
Hygromycin B 100 mg/ml in water (from Streptomyces hygroscopicus) , 90% (by HPLC), sterile filtered
Catalog # ENZOALX380309G001
Anbieter:  Enzo Life Sciences
CAS-Nummer:  
Hygromycin B 100 mg/ml in water (from Streptomyces hygroscopicus) , 90% (by HPLC), sterile filtered
Catalog # ENZOALX380309G001
Anbieter:  Enzo Life Sciences
CAS-Nummer:  

Technische Daten

  • Pk:
    1 g
  • Seq:
    3047

Technische Daten

Über diesen Artikel

 :
 :527.5 g/mol
Lagertemperatur:Refrigerator
CAS-Nummer:31282-04-9
Formel:C₂₀H₃₇N₃O₁₃
MDL-Nummer:MFCD06795479