

Vitamin A acetate (synthetic) ≥95% (by HPLC)
Supplier: ENZO LIFE SCIENCES
Certificates
About this item
CAS Number:127-47-9
:
Formula:C₂₂H₃₂O₂
MDL Number:MFCD00019413
:328.49 g/mol
EINECS:204-844-2
:GHS07
:GHS08
Storage Temperature:Refrigerator
Specification Test Results
- Purity (HPLC):≥95%
- Source:Synthetic
Specification Test Results
Frequently Bought Together