Auftragserfassung
Switzerland
ContactUsLinkComponent
ODN M362 (synthetisch), TLRgrade™
ODN M362 (synthetic), TLRgrade™
Catalog # ENZOALX746054M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
ODN M362 (synthetic), TLRgrade™
Catalog # ENZOALX746054M001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Source:
    Synthetic
  • Formulation:
    Lyophilised. Sterile.
  • Purity:
    Activity and endotoxin tested - TLRgrade™
  • Endotoxin content:
    <0.002 EU/µg

Specification Test Results

About this item

 :
 :8.073 g/mol
Storage Temperature:Refrigerator
Storage Temperature:Kühlschrank