ODN 1585 (synthetic), TLRgrade™
Catalog # ENZOALX746003C100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 µG
- Seq:1286
Specifications
Specification Test Results
- Source:Synthetic
- Formulation:Lyophilised. Sterile.
- Purity:Activity and endotoxin tested - TLRgrade™
- Endotoxin content:<0.002 EU/µg
- Quantity:14.9 nmol (~99µg)
Specification Test Results
About this item
:
:6.643 g/mol
Storage Temperature:Refrigerator