Order Entry
Switzerland
ContactUsLinkComponent
DAF-FM diacetate 2.5 mg/ml ≥98% (by HPLC)
Irritant
DAF-FM diacetate 2.5 mg/ml ≥98% (by HPLC)
Catalog # ENZOALX620071M001
Supplier:  Enzo Life Sciences
CAS Number:  
Irritant
DAF-FM diacetate 2.5 mg/ml ≥98% (by HPLC)
Catalog # ENZOALX620071M001
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    1 mg
  • Seq:
    2070

Specifications

Specification Test Results

  • Formulation:
    Dissolved in 0.4 ml DMSO
  • Purity:
    ≥98% (HPLC)

Specification Test Results

About this item

 :
 :496.4 g/mol
EINECS:200-664-3
 :GHS07
Storage Temperature:Freezer
CAS Number:254109-22-3
Formula:C₂₅H₁₈F₂N₂O₇