Order Entry
Switzerland
ContactUsLinkComponent
2',7'-Dichlorofluorescin diacetate ≥95% (by HPLC)
Irritant
2',7'-Dichlorofluorescin diacetate ≥95% (by HPLC)
Catalog # ENZOALX610022M050
Supplier:  Enzo Life Sciences
CAS Number:  
undefined
Irritant
2',7'-Dichlorofluorescin diacetate ≥95% (by HPLC)
Catalog # ENZOALX610022M050
Supplier:  Enzo Life Sciences
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

Specification Test Results

  • Purity:
    ≥95% (HPLC)

Specification Test Results

About this item

 :
 :487.285 g/mol
 :GHS07
Storage Temperature:Freezer
CAS Number:4091-99-0
Formula:C₂₄H₁₆Cl₂O₇
MDL Number:MFCD00128955