
Fenobam ≥98% (by 1H-NMR)
Katalognr. ENZOALX550413M005
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:5 mg
- Seq:2210
Specifications
Specification Test Results
- Purity (¹H-NMR):≥98%
- Identity:Determined by NMR
Specification Test Results
About this item
:
:266.7 g/mol
Storage Temperature:Refrigerator
UN:2811
ADR:6.1,III
:GHS06
CAS Number:57653-26-6
Formula:C₁₁H₁₁ClN₄O₂
MDL Number:MFCD00868019