Order Entry
Switzerland
ContactUsLinkComponent
Amiloride hydrochloride dihydrate ≥98%
Amiloride hydrochloride dihydrate ≥98%
Catalog # ENZOALX550212G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Amiloride hydrochloride dihydrate ≥98%
Catalog # ENZOALX550212G001
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :302.116 g/mol
CAS Number:17440-83-4
 :2609-46-3
Formula:C₆H₈ClN₇O·HCl·2H₂O
MDL Number:MFCD00058197