Sodium chloride 0.9% physiological saline endotoxin-free, sterile
Catalog # ENZOALX505009LD15
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1,5 mL
- Seq:2095
Specifications
Specification Test Results
- Endotoxin content:<0.0002 EU/µg (pyrotell gel-clot endotoxin test)
Specification Test Results
About this item
:
:58.44 g/mol
Storage Temperature:Refrigerator
CAS Number:7647-14-5
Formula:NaCl
MDL Number:MFCD00003477