
N-Octyl-β-D-glucopyranoside
Supplier: Enzo Life Sciences
Certificates
About this item
CAS Number:29836-26-8
:
Formula:C₁₄H₂₈O₆
MDL Number:MFCD00063288
:292.37 g/mol
Melting Pt:110 °C
EINECS:249-887-8
:GHS07
Storage Temperature:Freezer
Frequently Bought Together