Order Entry
Switzerland
ContactUsLinkComponent
 
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) ≥98% (by HPLC)
Catalog # ENZOALX480014G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) ≥98% (by HPLC)
Catalog # ENZOALX480014G005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    5 g
  • Seq:
    3173

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%

Specification Test Results

About this item

 :
 :391.2 g/mol
Merck Index:14,00158
Storage Temperature:Freezer
EINECS:224-961-2
CAS Number:4578-31-8
Formula:C₁₀H₁₂N₅Na₂O₇P
MDL Number:MFCD00065023