Order Entry
Switzerland
ContactUsLinkComponent
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC)
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC)
Catalog # ENZOALX480011M100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Adenosine-3',5'-cyclic monophosphate sodium salt ≥98% (by HPLC)
Catalog # ENZOALX480011M100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

Specification Test Results

  • Purity (HPLC):
    ≥98%
  • Identity:
    Identity determined by ³¹P- NMR, ¹H-NMR, FAB-MS and UV

Specification Test Results

About this item

 :
 :351.19 g/mol
Merck Index:14,02708
Storage Temperature:Freezer
CAS Number:37839-81-9
Formula:C₁₀H₁₁N₅NaO₆P
MDL Number:MFCD00069736