Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium butyrate
Sodium butyrate
Sodium butyrate
ENZOALX270301G001
:
ENZO LIFE SCIENCES
:
Sodium butyrate
ENZOALX270301G001
:
ENZO LIFE SCIENCES
:
Pack type:
Glass bottle for solids
Pk:
1 g
Seq:
3047
:
:
110.09 g/mol
:
244 °C
:
1,324 g/cm³ (30 °C)
:
205-857-6
:
156-54-7
:
C₄H₇NaO₂
:
MFCD00002816