Order Entry
Switzerland
ContactUsLinkComponent
AUDA ≥98% (by TLC)
AUDA ≥98% (by TLC)
Catalog # ENZOBMLEI3920050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
AUDA ≥98% (by TLC)
Catalog # ENZOBMLEI3920050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :392.575 g/mol
Storage Temperature:Freezer
CAS Number:479413-70-2
Formula:C₂₁H₃₆N₂O₃
MDL Number:MFCD12912267