Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Dorsomorphin ≥98% (by TLC)

Dorsomorphin ≥98% (by TLC)

Supplier:   Enzo Life Sciences
Dorsomorphin ≥98% (by TLC)
Supplier:  Enzo Life Sciences
Certificates
Dorsomorphin ≥98% (by TLC)
Dorsomorphin ≥98% (by TLC)

2 Options Available Below

Product Details & Documents

Cell-permeable pyrrazolopyrimidine derivative that inhibits AMP kinase (Ki=109nM in the absence of AMP) in an ATP-competitive manner. It displays no significant inhibition of ZAPK, SYK, PKCT, PKA and JAK3. Decreases food intake in mice and inhibits the effects of AICAR and metformin. It has been shown to inhibit BMP type I receptors ALK2, ALK3 and ALK6. Promotes cardiomyogenesis in mouse embryonic stem cells. Induces protective autophagy in cancer cell lines.

  • AMPK inhibitor

Yellow solid.

Soluble in DMSO (3mg/ml, warm)

Synonyms:  6-(4-(2-(Piperidin-1-yl)ethoxy)phenyl)-3-(pyridin-4-yl)pyrazolo[1,5-a]pyrimidine, 6-[4-(2-Piperidin-1-ylethoxy)phenyl]-3-pyridin-4-ylpyrazolo[1,5-a]pyrimidine
  • CAS Number:866405-64-3
  • Formula:C₂₄H₂₅N₅O
  • MDL Number:MFCD08705402
  • Molecular Weight:399.5 g/mol
  • Storage Temperature:Freezer
Product Family Options

Product Information

  • Pk
    Availability
    Price
  • 5 mg
    Log in for availability
    Log in for availability
    1 * 5 mg
    1 * 5 mg - 135.00 SFr.
    135.00 SFr.
    1 * 5 mg
       135.00 SFr.

  • 25 mg
    Log in for availability
    Log in for availability
    1 * 25 mg
    1 * 25 mg - 567.00 SFr.
    567.00 SFr.
    1 * 25 mg
       567.00 SFr.

Recommandations

Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".

Sinon, vous recevrez des recommandations génériques.