SBHA ≥95% (by TLC)
Catalog # ENZOBMLGR3230100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:204.2 g/mol
Storage Temperature:Ambient
CAS Number:38937-66-5
Formula:C₈H₁₆N₂O₄
MDL Number:MFCD00192455