Order Entry
Switzerland
ContactUsLinkComponent
Selumetinib ≥95% (by HPLC)
Selumetinib ≥95% (by HPLC)
Catalog # ENZOENZCHM1840050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Selumetinib ≥95% (by HPLC)
Catalog # ENZOENZCHM1840050
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    50 mg
  • Seq:
    2370

Specifications

About this item

 :
 :457.7 g/mol
CAS Number:606143-52-6
Formula:C₁₇H₁₅BrClFN₄O₃
MDL Number:MFCD11977472