Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Wedelolactone
Wedelolactone ≥98% (by TLC)
Wedelolactone ≥98% (by TLC)
Wedelolactone ≥98% (by TLC)
Catalog # ENZOBMLEI3160001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Wedelolactone ≥98% (by TLC)
Catalog # ENZOBMLEI3160001
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
314.2 g/mol
CAS Number:
524-12-9
Formula:
C₁₆H₁₀O₇
MDL Number:
MFCD07778564
Product Details