Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
TCPOBOP
TCPOBOP ≥98% (by TLC)
TCPOBOP ≥98% (by TLC)
ENZOBMLGR2450025
:
ENZO LIFE SCIENCES
:
TCPOBOP ≥98% (by TLC)
ENZOBMLGR2450025
:
ENZO LIFE SCIENCES
:
Pk:
25 mg
Seq:
2315
Source:
Synthetic
Purity:
≥98% (TLC)
:
:
402.1 g/mol
:
Ambient
:
76150-91-9
:
C₁₆H₈Cl₄N₂O₂
:
MFCD00057368