Pravadoline ≥98% (by TLC)
Catalog # ENZOBMLCR1100025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
Specification Test Results
- Purity:≥98% (TLC)
- Identity:Determined by NMR
Specification Test Results
About this item
:
:378.5 g/mol
Storage Temperature:Freezer
CAS Number:92623-83-1
Formula:C₂₃H₂₆N₂O₃