Order Entry
Switzerland
ContactUsLinkComponent
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95% (by HPLC), Ultrapure
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52303
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
3,3'-Di-n-hexyloxacarbocyanine iodide [DiOC6(3) iodide] ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52303
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    100 mg
  • Seq:
    2440

Specifications

About this item

 :
 :572.531 g/mol
Storage Temperature:Freezer
CAS Number:53213-82-4
Formula:C₂₉H₃₇IN₂O₂
MDL Number:MFCD00011954