N-Arachidonoyldopamine ≥98% (by TLC)
Catalog # ENZOBMLVR1040025
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Pk:25 mg
- Seq:2315
Specifications
About this item
:
:439.64 g/mol
CAS Number:199875-69-9
Formula:C₂₈H₄₁NO₃
MDL Number:MFCD03412031