Order Entry
Switzerland
ContactUsLinkComponent
Bromobimane (mBBr) ≥95% (by HPLC), Ultrapure
Bromobimane (mBBr) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52501
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
undefined
Bromobimane (mBBr) ≥95% (by HPLC), Ultrapure
Catalog # ENZOENZ52501
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Pk:
    25 mg
  • Seq:
    2315

Specifications

About this item

 :
 :271.11 g/mol
Storage Temperature:Freezer
CAS Number:71418-44-5
Formula:C₁₀H₁₁BrN₂O₂
MDL Number:MFCD00036951