Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Baicalein
Baicalein ≥98% (by TLC)
Baicalein ≥98% (by TLC)
Baicalein ≥98% (by TLC)
Catalog # ENZOBMLEI1060050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Expand image
Baicalein ≥98% (by TLC)
Catalog # ENZOBMLEI1060050
Supplier:
ENZO LIFE SCIENCES
CAS Number:
Specifications
Pk:
50 mg
Seq:
2370
Specifications
About this item
Product Details
:
:
270.24 g/mol
CAS Number:
491-67-8
Formula:
C₁₅H₁₀O₅
MDL Number:
MFCD00017459
Product Details