Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Absorbance 96 Plate Reader

Absorbance 96 Plate Reader

Absorbance 96 plate reader, 2,5 W, 405, 450, 492, 620 nm
Catalog # ENZOENZINSA96
Supplier:   Enzo Life Sciences
Absorbance 96 Plate Reader
Catalog # ENZOENZINSA96
Supplier:  Enzo Life Sciences
Absorbance 96 Plate Reader

Absorbance 96 plate reader, 2,5 W, 405, 450, 492, 620 nm

Price
SFr. 6’370.00
Log in for availability
1 * 1 ST
Product Details & Documents

The Absorbance 96 is a uniquely designed microplate reader for assays in 96-well format. By far the most compact reader on the market, the footprint of the plate reader is almost as small as the plate itself.

  • Space saving design
  • Fast readout speed
  • Reliable measurement results
  • Maintenance-free
  • Solid state technology

The Absorbance 96 delivers precise, accurate results at an affordable price. Together with its proprietary software, the Absorbance 96 is ideal for a variety of applications extending from ELISA to protein- and cell-based assays.

Connectivity/communication: Through USB connection 5 V
Detection method: Absorbance
Shipping: Ambient temperature

Caution: Research use only.

Specifications
  • Wavelength:
    405, 450, 492, 620 nm
  • Description:
    Absorbance 96 plate reader, 2,5 W
  • Cat. No.:
    ENZOENZINSA96
  • Light source:
    4×LEDs
  • Accuracy:
    405 nm: ≤1,5% + 0,010 OD (0 - 2 OD);≤3% + 0,010 OD (2 - 3 OD)/≥450 nm: ≤1% + 0,010 OD (0 - 2 OD)≤1,5% + 0,010 OD (2 - 3 OD)
  • Detector:
    Endpoint, kinetic (Detection mode)
  • Photometric range:
    0 - 4 OD
  • Reproducibility:
    ≤0,5% + 0,005 OD from 0,0 - 2,0 OD≤1,0% + 0,010 OD from 2,0 - 3,0 OD
  • Linearity:
    405 nm: ≤1,5% (0 - 2 OD); ≤3,0% (2 - 3 OD)/≥450 nm: ≤1% (0 - 2 OD); ≤1,5% (2 - 3 OD)
  • Voltage:
    5 V
  • Dimensions:
    154L×96W×55H mm
  • Software:
    Compatible with microsoft windows and MacOS
  • Weight:
    0,9 kg
  • Resolution:
    0,001 OD
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.