



Hygromycin B solution in PBS, sterile filtered
Catalog # ENZOALX380306G001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:527.5 g/mol
Storage Temperature:Refrigerator
EINECS:250-545-5
UN:2810
ADR:6.1,I
:GHS05
:GHS06
:GHS07
:GHS08
CAS Number:31282-04-9
Formula:C₂₀H₃₇N₃O₁₃
MDL Number:MFCD06795479