Stainless Steel Solid Cupboard Shelves
Supplier: TEKNOMEK
New Product
Certificates
About this item
Our stainless steel shelves are perfect for use with our range of cupboards, allowing you to customise your cupboard to your exact specification. Manufactured in 304L-grade stainless steel shelves for optimal sturdiness and durability.
- Brushed dull polish finish
- Available in solid
- Specifically for our cupboard range
Long-lasting and sturdy, the shelves have adjustment points every 180 mm, ideal for your storage requirements. The shelves are strong and versatile, offering you the perfect solution to space-efficient storage. Our stainless steel shelves are suited to rigorous hygiene and HACCP routines, meaning you can rely on us to deliver a hygienic, durable and easy to clean product ideal for your workspace.
Frequently Bought Together