Order Entry
Switzerland
ContactUsLinkComponent
SPC® Oil-only Absorbents
SPC® Oil-only Absorbents
Catalog # BRDYSKO-1100
Supplier:  Brady
SPC® Oil-only Absorbents
Catalog # BRDYSKO-1100
Supplier:  Brady

Specifications

  • Weight:
    121 kg
  • Description:
    Oil-only sorbent, 1000 litre container kit
  • W×D×H:
    1205×1085×1315 mm
  • Absorbency:
    972 L
  • Pk:
    1 KIT

Specifications

About this item

Drop front spill kit designed for larger spills of oil and petroleum based liquids. Top and drop front lid provides easy access to the absorbents inside the kit.

  • Absorbs up to 972 litres (full content of an IBC)
  • Absorbents and spill control
  • Drop front for fast and easy access

Wheeled to easily manoeuvre to location of the spill.

Delivery: Includes: 300 ENV200-M pads, 2 OP15-E rolls, 4 ENV510 booms, 30 OIL430 socs, 10 OIL 124 socs, 20 OIL1818 pillows, 1 pair SPK-HANDSCH gloves, 1 pair SPK-BRIL goggles, 50 disposal bags.