Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Fittings and Connectors
Cadence Micro-Mate Threaded End Adapter
Cadence Micro-Mate Threaded End Adapter
Cadence Micro-Mate Threaded End Adapter
Catalog # POPR6261IND
Supplier:
CADENCE SCIENCE
Bild vergrößern
Cadence Micro-Mate Threaded End Adapter
Catalog # POPR6261IND
Supplier:
CADENCE SCIENCE
Specifications
Thread:
0,125" NPT
Description:
FLL to M5 × 6 mm thread (plated brass)
Material:
Stainless steel
Pk:
1
Specifications
About this item
Product Details
Product Details