ContactUsLinkComponent
Freezer Racks for Nordic Lab ULT U250 Freezer
Freezer Racks for Nordic Lab ULT U250 Freezer
Catalog # TENATE23722
Supplier:  TENAK
Freezer Racks for Nordic Lab ULT U250 Freezer
Catalog # TENATE23722
Supplier:  TENAK
Specifications
  • Description:
    Standard rack, 10 boxes per rack
  • Pk:
    1
Specifications
About this item
Standard rack for the Nordic Lab ULT U250 freezer, in 2 different sizes.
  • No of racks: 12
  • No of boxes: 120