Order Entry
Switzerland
ContactUsLinkComponent
 
Organic Reference Standard, Vinylchloride 100 ug/ml in Methanol
Catalog # CMPLP857620
CAS Number:  
Organic Reference Standard, Vinylchloride 100 ug/ml in Methanol
Catalog # CMPLP857620
CAS Number:  

Specifications

  • Pk:
    1 mL
  • Description:
    Vinylchloride 100 ug/ml in Methanol
  • Storage conditions:
    Store in a freezer at −18 °C or below
  • GHS signal:
    Danger
  • Melting point:
    −98 °C
  • Shelf life:
    24 months
  • Density:
    0,79 g/cm³ at 20 °C
  • Flash point:
    11 °C
  • Boiling point:
    64 °C
  • Volume:
    1 ml

Specifications

About this item