VISOCOLOR® Colorimetric Test, Zinc
Supplier: MACHEREY-NAGEL
Certificates
About this item
Reagents for the determination of zinc in water samples.
- Combines simple implementation with high measurement reliability
Simple determination directly on site or in the laboratory.
Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on PF-12Plus. Range on other photometers can be different
Specifications
- Modell:MANA931098
- Platform:Colorimetric test kits
- Lagertemperatur:15...25 °C
- Haltbarkeit (Monate):12
- Gewicht:234,8 g (Including packaging)