ContactUsLinkComponent
Gem Runner™ Reloadable Single Edge Blade Safety Dispenser
Gem Runner™ Reloadable Single Edge Blade Safety Dispenser
Catalog # PERSAGTL-7000-0000
Supplier:  PERSONNA MEDICAL
Gem Runner™ Reloadable Single Edge Blade Safety Dispenser
Catalog # PERSAGTL-7000-0000
Supplier:  PERSONNA MEDICAL
Specifications
  • Description:
    Gem® Reloadable safety dispenser, plastic/metal
  • Packed:
    5 dispensers/case
  • Pk:
    5
Specifications
About this item
Safety dispenser fits 0,009 and 0,012" single edge blades.
  • Great for high volume uses
  • Blades not included
Holds all Gem®3 and Gem®2 cartridges.
Cartridges
Product Information