PMI Stock Standard A
Catalog # ACCUM-1666A-SSA-R2
Supplier: AccuStandard
CAS Number:
Specifications
- Pk:1 mL
- Description:PMI Stock standard A with 7 number of components
Specifications
About this item
PMI Stock standard A at stated concentration (µg/ml) in water with seven components.
- 1-Butanol - 2500 µg/ml
- t-Butanol - 2500 µg/ml
- 2-Furaldehyde - 2500 µg/ml
- Isobutyraldehyde - 2500 µg/ml
- Isopropanol - 1000 µg/ml
- 4-Methyl-2-pentanone - 1000 µg/ml
- 1-Pentanol - 2500 µg/ml
Storage condition: Keep refrigerated at 0...5 °C
Melting point: 0 °C (32 °F)
Boiling point: 100 °C (212 °F)