Order Entry
Switzerland
ContactUsLinkComponent
Trichloroacetic acid (TCA) 4% in aqueous solution Reag. Ph. Eur.
Corrosive
Irritant
Environment
Trichloroacetic acid (TCA) 4% in aqueous solution Reag. Ph. Eur.
Catalog # 87956.180
Supplier:  VWR Chemicals
CAS Number:  
undefined
Corrosive
Irritant
Environment
Trichloroacetic acid (TCA) 4% in aqueous solution Reag. Ph. Eur.
Catalog # 87956.180
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

About this item

 :
 :163.39 g/mol
Storage Temperature:Ambient
EINECS:200-927-2
UN:2564
ADR:8,III
 :GHS05
 :GHS07
 :GHS09
CAS Number:76-03-9
Formula:Cl₃CCOOH
MDL Number:MFCD00004177