

Hydrochloric acid 25% Reag. Ph. Eur. 1043501
Catalog # 87842.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.13 g/cm³ (20 °C)
ADR:8,II
:36.46 g/mol
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
REACH:01-2119484862-27
:GHS05
:GHS07
CAS Number:7647-01-0
Formula:HCl
MDL Number:MFCD00011324