Barium hydroxide 0.15 mol/l (0.3 N) in aqueous solution Reag. Ph. Eur. 1009401
Catalog # 87785.290
Supplier: VWR Chemicals
CAS Number:
Specifications
- Pack type:Plastic bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:171.34 g/mol
EINECS:241-234-5
CAS Number:17194-00-2
Formula:H₂BaO₂
MDL Number:MFCD00003443