Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

5α-Cholestane

Supplier:   MATREYA
5α-Cholestane
Supplier:  MATREYA
Certificates

1 Options Available Below

Product Details & Documents
  • CAS Number:481-21-0
  • Formula:C₂₇H₄₈
  • MDL Number:MFCD00066412
  • Molecular Weight:372.68 g/mol
  • Merck Index:12,02254
  • Boiling Pt:441 °C (1013 hPa)
  • Melting Pt:79 °C
  • Density:0.9133 g/cm³ (80 °C)
  •  :Density: 0.9133 g/cm³ (80 °C)
  • EINECS:207-562-8
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    100 mg
    Log in for availability
    Log in for availability
    1 * 100 mg
    1 * 100 mg

  • Glass bottle for solids
    100 mg
    Discontinued
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.