Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

5α-Cholestane 98+%

5α-Cholestane 98+%
Certificates
5α-Cholestane 98+%

3 Options Available Below

Product Details & Documents
  • CAS Number:481-21-0
  • Formula:C₂₇H₄₈
  • MDL Number:MFCD00066412
  • Molecular Weight:372.68 g/mol
  • Merck Index:12,02254
  • Boiling Pt:250 °C (1 mmHg)
  • Melting Pt:78…81 °C
  • Density:0,9133 g/cm³ (80 °C)
  •  :Density: 0.9133 g/cm³ (80 °C)
  • Storage Temperature:Ambient
  • EINECS:207-562-8

 

Specifications for Product Family

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    78 to 81 °C
  • GC:
    ≥98.0 %
  • Specific optical rotation:
    +23° to +30° (20 °C, 589 nm) (c=2, CHCl3)
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    250 mg
    Log in for availability
    Log in for availability
    1 * 250 mg
    1 * 250 mg - SFr. 111.00
    SFr. 111.00
    1 * 250 mg
       SFr. 111.00

  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - SFr. 300.00
    SFr. 300.00
    1 * 1 g
       SFr. 300.00

  • Glass bottle
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - SFr. 1’040.00
    SFr. 1’040.00
    1 * 5 g
       SFr. 1’040.00