Cholesterol
Supplier: MATREYA
Jump to:
About this item
Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
- CAS Number:57-88-5
- Formula:C₂₇H₄₆O
- MDL Number:MFCD00003646
- Molecular Weight:386.66 g/mol
- Merck Index:13,02221
- Boiling Pt:360 °C (1013 hPa)
- Melting Pt:147…150 °C
- Density:1.07 g/cm³ (20 °C)
- Storage Temperature:Freezer
- EINECS:200-353-2
1 Options Available Below
Product Details & Documents
Synonyms: Cholest-5-en-3-ol, Cholesterin, (3Beta)-Cholest-5-en-3-ol
- CAS Number:57-88-5
- Formula:C₂₇H₄₆O
- MDL Number:MFCD00003646
- Molecular Weight:386.66 g/mol
- Merck Index:13,02221
- Boiling Pt:360 °C (1013 hPa)
- Melting Pt:147…150 °C
- Density:1.07 g/cm³ (20 °C)
- Storage Temperature:Freezer
- EINECS:200-353-2
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



