Order Entry
Switzerland
Orders LinkContactUsLinkComponent
β-Alanine

β-Alanine

Glass bottle for solids, 1 kg
Catalog # MOLEM12123234
Marketsource
Supplier:   Molekula
β-Alanine
Catalog # MOLEM12123234
Supplier:  Molekula
Marketsource
CAS Number:  107-95-9
β-Alanine

Glass bottle for solids, 1 kg

CAS Number:  107-95-9
Price
Inquire For Price
Log in for availability
1 * 1 kg
Product Details & Documents
Synonyms:  H-β-Ala-OH, 3-Aminopropionic acid, H-Bala-OH, 3-Aminopropanoic acid
  • CAS Number:107-95-9
  • Formula:C₃H₇NO₂
  • MDL Number:MFCD00008200
  • Molecular Weight:89.09 g/mol
  • Merck Index:13,00204
  • Boiling Pt:201 °C (1013 hPa)
  • Melting Pt:94 °C
  • Density:1.44 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:203-536-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    1 kg

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.